Tisk…

Diethylether Stabilised

Dodavatel: Lach-ner S.R.O.
Nebezpečí

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

20018AT0M5000 20018AT0M2500 20018AT0M1000
LACH20018AT0M5000EA 0 CZK
LACH20018AT0M5000 LACH20018AT0M2500 LACH20018AT0M1000
Diethylether Stabilised
Diethylether
Vzorec: (CH₃CH₂)₂O
MW: 74.12
Bod varu: 35
Bod tání: –123 °C
Hustota: 0.7135 g/cm³ (20 °C)
Bod vzplanutí: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS číslo: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR