Tisk…
Bottle-top dispensers, Finnpipette™
Dodavatel: Thermo Fisher Scientific
4421120 4421130 4421140 4421150 4421160 4421170
LABS4421120EA 13060 CZK
LABS4421120 LABS4421130 LABS4421140 LABS4421150 LABS4421160 LABS4421170
Bottle-top dispensers, Finnpipette™
Dávkovače na lahve
This bottle top dispenser feature a superior tool for dosing liquids from reagent bottles that have six models available.
  • Ergonomically designed, non-slip piston handle
  • Excellent chemical and thermal resistance
  • Volumes are absolutely reproducible
  • Threaded suction hose for bubble free suction
They can be fully autoclaved without disassembling and have precise self-locking system in the volume setting.