Tisk…
SkanIt™ Upgrade package, version 5.0
Dodavatel: THERMO LABSYSTEMS LIFE SCIENCE
N17646 N18600
LABLN17646EA 94910 CZK
LABLN17646 LABLN18600
SkanIt™ Upgrade package, version 5.0
Software Laboratory Software
This SkanIt Software upgrade package is specifically designed for Thermo Scientific Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers to enable the use of SkanIt software and USB connection between instrument and PC. The upgrade package is also compatible with the discontinued Fluoroskan Ascent, Fluoroskan Ascent FL, and Luminoskan Ascent microplate readers. SkanIt Software is available in two editions: a research edition for scientists working in life science research and a drug discovery edition with features for compliancy with FDA 21 CFR Part 11. Note: the upgrade package isMore Product Information