Tisk…

3-(Phenylsulphonyl)propionitrile 99%

Dodavatel: Thermo Fisher Scientific
Varování

L20196.06 L20196.14
L20196.06EA 1380 CZK
L20196.06 L20196.14
3-(Phenylsulphonyl)propionitrile 99%
3-(Phenylsulphonyl)propionitrile

Purity: 99% CAS No.: 10154-75-3 Molecular Formula: C9H9NO2S Formula Weight: 195.24 Odour: Odourless Melting Point: 94-96° Beilstein Reference: 2051019 MDL No.: MFCD00047798

Vzorec: C₉H₉NO₂S
MW: 195.24 g/mol
Bod tání: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00047798
CAS číslo: 10154-75-3
EINECS: 233-424-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR