Tisk…

4-Bromo-2-fluorobenzoyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L20142.03 L20142.06 L20142.14
L20142.03EA 1140 CZK
L20142.03 L20142.06 L20142.14
4-Bromo-2-fluorobenzoyl chloride ≥98%
4-Bromo-2-fluorobenzoyl chloride

Purity: 98% CAS No.: 151982-51-3 Molecular Formula: C7H3BrClFO Formula Weight: 237.45 Refractive Index: 1.5877 Melting Point: 24-25° Boiling Point: 86-88°/15mm Beilstein Reference: 6323925 UN No.: 3261 MDL No.: MFCD03411584

Vzorec: C₇H₃BrClFO
Bod varu: 86…88 °C (15 mmHg)
Bod tání: 24…25 °C
Storage Temperature: Ambient
MDL Number: MFCD03411584
CAS číslo: 151982-51-3
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR