Tisk…

6-(Trifluoromethyl)nicotinic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L20054.03 L20054.MD
L20054.03EA 3200 CZK
L20054.03 L20054.MD
6-(Trifluoromethyl)nicotinic acid ≥97%
6-(Trifluoromethyl)nicotinic acid

Purity: 97% CAS No.: 231291-22-8 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Melting Point: 191-192° MDL No.: MFCD00792430

Vzorec: C₇H₄F₃NO₂
MW: 191.11 g/mol
Bod tání: 191…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00792430
CAS číslo: 231291-22-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR