Tisk…

3-Chloro-2-hydroxypyridine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L20027.03 L20027.06 L20027.14
L20027.03EA 889 CZK
L20027.03 L20027.06 L20027.14
3-Chloro-2-hydroxypyridine ≥97%
3-Chloro-2-hydroxypyridine

Purity: 97% CAS No.: 13466-35-8 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 180-182° Beilstein Reference: 109845

Vzorec: C₅H₄ClNO
Bod tání: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD01646109
CAS číslo: 13466-35-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR