Tisk…

2,6-Dichloronicotinic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L20002.03 L20002.06
L20002.03EA 1220 CZK
L20002.03 L20002.06
2,6-Dichloronicotinic acid ≥97%
2,6-Dichloronicotinic acid

Purity: 97% CAS No.: 38496-18-3 Molecular Formula: C6H3Cl2NO2 Formula Weight: 192.00 Melting Point: 146-151° Beilstein Reference: 136114

Vzorec: C₆H₃Cl₂NO₂
Bod tání: 146…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00075583
CAS číslo: 38496-18-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR