Tisk…

Boc-Thz-OH 98%

Dodavatel: Thermo Fisher Scientific

L19488.03 L19488.06
L19488.03EA 1470 CZK
L19488.03 L19488.06
Boc-Thz-OH 98%
Boc-Thz-OH

Purity: 98% CAS No.: 51077-16-8 Molecular Formula: C9H15NO4S Formula Weight: 233.28 Melting Point: ca 133° dec. Beilstein Reference: 6686084 Optical Rotation: -113° (c=1 in methanol) MDL No.: MFCD00077002

Vzorec: C₉H₁₅NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00077002
CAS číslo: 51077-16-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR