Tisk…

(R)-(-)-Mandelic acid ≥99%, ee 99%, ChiPros®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L19116.14 L19116.06
L19116.14EA 2900 CZK
L19116.14 L19116.06
(R)-(-)-Mandelic acid ≥99%, ee 99%, ChiPros®
(R)-(-)-Mandelic acid

Purity: 99+% CAS No.: 611-71-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 131-135° Beilstein Reference: 2691094

Vzorec: C₈H₈O₃
Bod varu: 322 °C (1013 hPa)
Bod tání: 131…135 °C
Hustota: 1,341 g/cm³ (20 °C)
Bod vzplanutí: >190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064251
CAS číslo: 611-71-2
EINECS: 210-276-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR