Tisk…

1-(Trifluoromethyl)cyclopropanecarboxylic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-(Trifluoromethyl)cyclopropane-1-carboxylic acid

L17957.03 L17957.06
L17957.03EA 4470 CZK
L17957.03 L17957.06
1-(Trifluoromethyl)cyclopropanecarboxylic acid ≥97%
1-(Trifluoromethyl)cyclopropanecarboxylic acid

Purity: 97% CAS No.: 277756-46-4 Molecular Formula: C5H5F3O2 Formula Weight: 154.09 Melting Point: 87-89° Boiling Point: 182-184° UN No.: 3261 MDL No.: MFCD03093070

Vzorec: C₅H₅F₃O₂
MW: 154.09 g/mol
Bod varu: 182…184 °C
Bod tání: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03093070
CAS číslo: 277756-46-4
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR