Tisk…

Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Diethyl [(ethoxycarbonyl)(fluoro)methyl]phosphonate, Triethyl 2-fluoro-2-phosphonoacetate

L17913.03 L17913.06 L17913.14
L17913.03EA 1470 CZK
L17913.03 L17913.06 L17913.14
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 96%
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent)

Purity: 96% CAS No.: 2356-16-3 Molecular Formula: C8H16FO5P Formula Weight: 242.19 Flash Point: 73°(163°F) Refractive Index: 1.4250 Boiling Point: 75°/0.01mm Beilstein Reference: 1912161 MDL No.: MFCD00134400

Vzorec: C₈H₁₆FO₅P
MW: 242.19 g/mol
Bod varu: 75 °C (0.01 mmHg)
Hustota: 1.194
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134400
CAS číslo: 2356-16-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR