Tisk…

DL-Valine ethyl ester hydrochloride 99%

Dodavatel: Thermo Fisher Scientific

L17906.14 L17906.06
L17906.14EA 4260 CZK
L17906.14 L17906.06
DL-Valine ethyl ester hydrochloride 99%
DL-Valine ethyl ester hydrochloride

Purity: 99% CAS No.: 23358-42-1 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Melting Point: ca 94° Beilstein Reference: 3912411 MDL No.: MFCD00067542

Vzorec: C₇H₁₅NO₂·HCl
Bod tání: ∼ 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00067542
CAS číslo: 23358-42-1
EINECS: 245-606-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR