Tisk…

2,4-Dichloro-6-iodoaniline ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L17659.06 L17659.14
L17659.06EA 1680 CZK
L17659.06 L17659.14
2,4-Dichloro-6-iodoaniline ≥97%
2,4-Dichloro-6-iodoaniline

Purity: 97% CAS No.: 697-90-5 Molecular Formula: C6H4Cl2IN Formula Weight: 287.92 Melting Point: 79-81° Beilstein Reference: 3246130 UN No.: 2811 MDL No.: MFCD00007661

Vzorec: C₆H₄Cl₂IN
Bod tání: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007661
CAS číslo: 697-90-5
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR