Tisk…

5-(tert-Butoxycarbonylamino)-1-pentanol 96%

Dodavatel: Thermo Fisher Scientific

L17649.03 L17649.06
L17649.03EA 1860 CZK
L17649.03 L17649.06
5-(tert-Butoxycarbonylamino)-1-pentanol 96%
5-(tert-Butoxycarbonylamino)-1-pentanol

Purity: 96% CAS No.: 75178-90-4 Molecular Formula: C10H21NO3 Formula Weight: 203.28 Storage Temperature: 2°C to 8°C Flash Point: 109°(228°F) Refractive Index: 1.4560 Beilstein Reference: 3603659 MDL No.: MFCD01862955

Vzorec: C₁₀H₂₁NO₃
Hustota: 1,000
Bod vzplanutí: >109 °C (228 °F)
MDL Number: MFCD01862955
CAS číslo: 75178-90-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR