Tisk…

Silikagel 99.9%, powder 0,5 µm

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

L16985.14 L16985.22
L16985.14EA 2820 CZK
L16985.14 L16985.22
Silikagel 99.9%, powder 0,5 µm
Silicon dioxide

Purity: 99.9% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Odour: Odourless Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Vzorec: SiO₂
Bod varu: 2230 °C
Bod tání: 1710 °C
Hustota: 2,0
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS číslo: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR