Tisk…

3-Aminophthalhydrazide monosodium salt ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L15205.03 L15205.06
L15205.03EA 2100 CZK
L15205.03 L15205.06
3-Aminophthalhydrazide monosodium salt ≥98%
3-Aminophthalhydrazide monosodium salt

Purity: 98+% CAS No.: 20666-12-0 Molecular Formula: C8H6N3NaO2 Formula Weight: 199.15 MDL No.: MFCD00161491

Vzorec: C₈H₆N₃NaO₂
Storage Temperature: Ambient
MDL Number: MFCD00161491
CAS číslo: 20666-12-0
EINECS: 629-344-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR