Tisk…

(2-Iodophenyl)acetic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-(2-Iodophenyl)acetic acid

L15116.06
L15116.06EA 1910 CZK
L15116.06
(2-Iodophenyl)acetic acid ≥98%
(2-Iodophenyl)acetic acid

Purity: 98+% CAS No.: 18698-96-9 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Melting Point: 115-117° Beilstein Reference: 2086800

Vzorec: (C₆H₄I)CH₂COOH
MW: 262,05 g/mol
Bod tání: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00046546
CAS číslo: 18698-96-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR