Tisk…

Akrylamid 99+% for electrophoresis

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L15075.22 L15075.36
L15075.22EA 1830 CZK
L15075.22 L15075.36
Akrylamid 99+% for electrophoresis
Akrylamid

Purity: 99+% CAS No.: 79-06-1 Molecular Formula: C3H5NO Formula Weight: 71.08 Flash Point: 138°(280°F) Odour: Odourless Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032

Vzorec: CH₂=CHCONH₂
Bod varu: 125 °C (25 mmHg)
Bod tání: 82…85 °C
Hustota: 1,322
Bod vzplanutí: 138 °C (280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008032
CAS číslo: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
Merck Index: 14,129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR