Tisk…

terc-Butylmethylether ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L14030.AP L14030.AU L14030.0F AB171832.2500
L14030.APEA 920 CZK
L14030.AP L14030.AU L14030.0F ABCRAB171832.2500
terc-Butylmethylether ≥99%
terc-Butylmethylether

Purity: 99% CAS No.: 1634-04-4 Molecular Formula: (CH3)3COCH3 Formula Weight: 88.15 Flash Point: -25°(-13°F) Refractive Index: 1.3690 Odour: Mild Melting Point: -109° Boiling Point: 55-56° Beilstein Reference: 1730942 Merck Reference: 14,6032 UN No.: 2350 MDL No.: MFCD00008812

Vzorec: (CH₃)₃COCH₃
Bod varu: 55…56 °C
Bod tání: –109 °C
Hustota: 0,742
Bod vzplanutí: –25 °C (–13 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS číslo: 1634-04-4
EINECS: 216-653-1
UN: 2350
ADR: 3,II
Merck Index: 14,6032
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR