Tisk…

3-Methylpiperidine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L13408.14 L13408.22
L13408.14EA 876 CZK
L13408.14 L13408.22
3-Methylpiperidine ≥98%
3-Methylpiperidine

Purity: 98% CAS No.: 626-56-2 Molecular Formula: C6H13N Formula Weight: 99.18 Flash Point: 8°(46°F) Refractive Index: 1.4470 Odour: Amine-like Melting Point: -24° Boiling Point: 124-125° Beilstein Reference: 79807 UN No.: 2733 MDL No.: MFCD00005994

Vzorec: C₆H₁₃N
MW: 99.18
Bod varu: 124…125 °C
Bod tání: –24 °C
Hustota: 0.8635
Bod vzplanutí: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005994
CAS číslo: 626-56-2
EINECS: 210-953-6
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR