Tisk…

Trimethyl orthoacetate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L13369.AD L13369.AK
L13369.ADEA 1250 CZK
L13369.AD L13369.AK
Trimethyl orthoacetate ≥98%
Trimethyl orthoacetate

Purity: 98% CAS No.: 1445-45-0 Molecular Formula: C5H12O3 Formula Weight: 120.15 Flash Point: 16°(60°F) Refractive Index: 1.3890 Boiling Point: 107-109° Beilstein Reference: 1098338 UN No.: 3272 MDL No.: MFCD00008477

Vzorec: C₅H₁₂O₃
Bod varu: 107…109 °C
Bod tání: –58 °C
Hustota: 0,956
Bod vzplanutí: 16 °C (60 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008477
CAS číslo: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR