Tisk…

Sodium bis(trimethylsilyl)amide 2 M in THF

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: HMDS sodium salt, N-Sodiohexamethyldisilazane, Sodium bis(trimethylsilyl)amide, HMDS-Na

L13352.AE L13352.AP
L13352.AEEA 2920 CZK
L13352.AE L13352.AP
Sodium bis(trimethylsilyl)amide 2 M in THF
Sodium bis(trimethylsilyl)amide

CAS No.: 1070-89-9 Molecular Formula: C6H18NNaSi2 Formula Weight: 183.38 Flash Point: -17°(1°F) Beilstein Reference: 3629917 UN No.: 2924 MDL No.: MFCD00009835

Vzorec: C₆H₁₈NNaSi₂
MW: 183.38
Hustota: 1.065
Bod vzplanutí: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009835
CAS číslo: 1070-89-9
EINECS: 213-983-8
UN: 2924
ADR: 3 II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR