Tisk…

6-Nitro-m-anisic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L13346.06 L13346.14
L13346.06EA 718 CZK
L13346.06 L13346.14
6-Nitro-m-anisic acid ≥97%
6-Nitro-m-anisic acid

Purity: 97% CAS No.: 1882-69-5 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 130-135° Beilstein Reference: 1877657

Vzorec: C₈H₇NO₅
Bod tání: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00151836
CAS číslo: 1882-69-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR