Tisk…

(4-Nitrophenyl)-β-D-galactopyranoside hydrate ≥98%

Dodavatel: Thermo Fisher Scientific

L12911.03 L12911.06
L12911.03EA 1680 CZK
L12911.03 L12911.06
(4-Nitrophenyl)-β-D-galactopyranoside hydrate ≥98%
(4-Nitrophenyl)-β-D-galactopyranoside hydrate

Purity: 98+% CAS No.: 200422-18-0 Molecular Formula: C12H15NO8.xH2O Formula Weight: 319.27 (301.25anhy) Storage Temperature: 2°C to 8°C Melting Point: 180-184° Beilstein Reference: 92213 Optical Rotation: -84° (c=0.3 in water) MDL No.: MFCD00150725

Vzorec: C₁₂H₁₅NO₈·H₂O
Bod tání: 180…184 °C
MDL Number: MFCD00150725
CAS číslo: 200422-18-0
EINECS: 221-584-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR