Tisk…

3-(Trifluoromethyl)phenylhydrazine 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-(3-(Trifluoromethyl)phenyl)hydrazine, 3-Hydrazinobenzotrifluoride

L11996.03 L11996.06
L11996.03EA 1910 CZK
L11996.03 L11996.06
3-(Trifluoromethyl)phenylhydrazine 95%
3-(Trifluoromethyl)phenylhydrazine

Purity: 95% CAS No.: 368-78-5 Molecular Formula: C7H7F3N2 Formula Weight: 176.14 Flash Point: 107°(224°F) Refractive Index: 1.5040 Boiling Point: 80-83°/9mm Beilstein Reference: 640075 MDL No.: MFCD00025093

Vzorec: C₇H₇F₃N₂
MW: 176.14
Bod varu: 80…83 °C (9 mmHg)
Hustota: 1.348
Bod vzplanutí: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00025093
CAS číslo: 368-78-5
EINECS: 206-713-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR