Tisk…

Uhlík aktivní, powder, Norit® GSX, washed with sulphuric acid, steam activated and acid washed

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Carbon activated, Activated carbon, Activated charcoal

L11860.22 L11860.36 L11860.0P L11860.A3
L11860.22EA 1040 CZK
L11860.22 L11860.36 L11860.0P L11860.A3
Uhlík aktivní, powder, Norit® GSX, washed with sulphuric acid, steam activated and acid washed
Uhlík aktivní

CAS No.: 7440-44-0 UN No.: 1362 MDL No.: MFCD00133992

Vzorec: C
MW: 12.01
Bod tání: 3550
Hustota: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS číslo: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2 III
REACH: 01-2119488894-16
Merck Index: 13,01818

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR