Tisk…

6-Hydroxy-3-coumaranone ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L10951.03 L10951.06
L10951.03EA 1120 CZK
L10951.03 L10951.06
6-Hydroxy-3-coumaranone ≥97%
6-Hydroxy-3-coumaranone

Purity: 97% CAS No.: 6272-26-0 Molecular Formula: C8H6O3 Formula Weight: 150.14 Melting Point: 240-242° Beilstein Reference: 121444

Vzorec: C₈H₆O₃
Bod tání: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00068174
CAS číslo: 6272-26-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR