Tisk…

Ethyl pentafluorobenzoate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L10523.06 L10523.14
L10523.06EA 954 CZK
L10523.06 L10523.14
Ethyl pentafluorobenzoate ≥98%
Ethyl pentafluorobenzoate

Purity: 98% CAS No.: 4522-93-4 Molecular Formula: C9H5F5O2 Formula Weight: 240.13 Flash Point: 80°(176°F) Refractive Index: 1.4280 Boiling Point: 112-114° Beilstein Reference: 2054770 MDL No.: MFCD00039211

Vzorec: C₉H₅F₅O₂
Bod varu: 112…114 °C
Hustota: 1,431
Bod vzplanutí: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039211
CAS číslo: 4522-93-4
EINECS: 224-854-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR