Tisk…

2,5-Thiophenedicarboxylic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Thiophene-2,5-dicarboxylic acid

L10293.03 L10293.06 L10293.14
L10293.03EA 893 CZK
L10293.03 L10293.06 L10293.14
2,5-Thiophenedicarboxylic acid ≥97%
2,5-Thiophenedicarboxylic acid

Purity: 97% CAS No.: 4282-31-9 Molecular Formula: C6H4O4S Formula Weight: 172.16 Melting Point: 355-360° Beilstein Reference: 136781

Vzorec: C₆H₄O₄S
MW: 172.16 g/mol
Bod tání: 355…360 °C
Storage Temperature: Ambient
MDL Number: MFCD00016896
CAS číslo: 4282-31-9
EINECS: 224-284-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR