Tisk…

(4-Nitrophenyl)-α-D-glucopyranoside ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 4-Nitrophenyl α-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl a-D-glucopyranoside

L10070.03 L10070.06
L10070.03EA 2460 CZK
L10070.03 L10070.06
(4-Nitrophenyl)-α-D-glucopyranoside ≥98%
(4-Nitrophenyl)-α-D-glucopyranoside

Purity: 98+% CAS No.: 3767-28-0 Molecular Formula: C12H15NO8 Formula Weight: 301.25 Storage Temperature: 2°C to 8°C Melting Point: 206-207° Beilstein Reference: 92212 Optical Rotation: +218° (c=0.5 in water) MDL No.: MFCD00064088

Vzorec: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Bod tání: 206…207 °C
MDL Number: MFCD00064088
CAS číslo: 3767-28-0
EINECS: 223-189-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR