Tisk…

DL-3-Phenyllactic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L09854.03 L09854.06
L09854.03EA 999 CZK
L09854.03 L09854.06
DL-3-Phenyllactic acid ≥98%
DL-3-Phenyllactic acid

Purity: 98+% CAS No.: 828-01-3 Molecular Formula: C9H10O3 Formula Weight: 166.18 Odour: Odourless Melting Point: 95-98° Beilstein Reference: 2209791 MDL No.: MFCD00065928

Vzorec: C₉H₁₀O₃
Bod tání: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00065928
CAS číslo: 828-01-3
EINECS: 212-580-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR