Tisk…

Methyl m-anisate ≥98%

Dodavatel: Thermo Fisher Scientific

L09041.09 L09041.18
L09041.09EA 1310 CZK
L09041.09 L09041.18
Methyl m-anisate ≥98%
Methyl m-anisate

Purity: 98% CAS No.: 5368-81-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Refractive Index: 1.5270 Boiling Point: 238° Beilstein Reference: 2208567 MDL No.: MFCD00017193

Vzorec: C₉H₁₀O₃
Bod varu: 238 °C
Hustota: 1,15
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017193
CAS číslo: 5368-81-0
EINECS: 226-359-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR