Tisk…

N-(tert-Butoxycarbonyl)-D-serine ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: N-Boc-D-serine, Boc-D-Ser-OH

L08904.03 L08904.06
L08904.03EA 1190 CZK
L08904.03 L08904.06
N-(tert-Butoxycarbonyl)-D-serine ≥98%
N-(tert-Butoxycarbonyl)-D-serine

Purity: 98+% CAS No.: 6368-20-3 Molecular Formula: C8H15NO5 Formula Weight: 205.21 Storage Temperature: 2°C to 8°C Melting Point: 88-91° Beilstein Reference: 1874714 Optical Rotation: +3.5° (c=2 in acetic acid) MDL No.: MFCD00063142

Vzorec: C₈H₁₅NO₅
MW: 205.21 g/mol
Bod tání: 88…91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063142
CAS číslo: 6368-20-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR