Tisk…

Dimethyl-5-norbornene-2,3-dicarboxylate 94%

Dodavatel: Thermo Fisher Scientific
Varování

L08664.09 L08664.18
L08664.09EA 2760 CZK
L08664.09 L08664.18
Dimethyl-5-norbornene-2,3-dicarboxylate 94%
Dimethyl-5-norbornene-2,3-dicarboxylate

Purity: 94% CAS No.: 5826-73-3 Molecular Formula: C11H14O4 Formula Weight: 210.23 Boiling Point: 150-152°/20mm Beilstein Reference: 1248588 MDL No.: MFCD00154455

Vzorec: C₁₁H₁₄O₄
Bod varu: 150…152 °C (20 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00154455
CAS číslo: 5826-73-3
Merck Index: 14,3240

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR