Tisk…

Potassium-2,5-dihydroxybenzenesulphonate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,5-Dihydroxybenzenesulphonic acid potassium sal, Hydroquinonesulphonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulphonate

L08406.22 L08406.36
L08406.22EA 1230 CZK
L08406.22 L08406.36
Potassium-2,5-dihydroxybenzenesulphonate ≥98%
Potassium-2,5-dihydroxybenzenesulphonate

Purity: 98+% CAS No.: 21799-87-1 Molecular Formula: C6H5KO5S Formula Weight: 228.27 Melting Point: ca 251° dec. Beilstein Reference: 5187027

Vzorec: C₆H₅KO₅S
Storage Temperature: Ambient
MDL Number: MFCD00007475
CAS číslo: 21799-87-1
EINECS: 244-584-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR