Tisk…

D-(-)-Norleucine ≥99%

Dodavatel: Thermo Fisher Scientific

L08257.03 L08257.06
L08257.03EA 1480 CZK
L08257.03 L08257.06
D-(-)-Norleucine ≥99%
D-(-)-Norleucine

Purity: 99% CAS No.: 327-56-0 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721749 Merck Reference: 14,6706 Optical Rotation: -23° (c=5 in 5N HCl) MDL No.: MFCD00008099

Vzorec: C₆H₁₃NO₂
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008099
CAS číslo: 327-56-0
EINECS: 206-320-9
Merck Index: 14,6706

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR