Tisk…

Triethyl orthopropionate ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L08172.AE L08172.AP
L08172.AEEA 1390 CZK
L08172.AE L08172.AP
Triethyl orthopropionate ≥97%
Triethyl orthopropionate

Purity: 98% CAS No.: 115-80-0 Molecular Formula: C9H20O3 Formula Weight: 176.26 Flash Point: 45°(113°F) Refractive Index: 1.4020 Odour: Slightly sweetish Boiling Point: 158° Beilstein Reference: 906798 UN No.: 3272 MDL No.: MFCD00009226

Vzorec: C₉H₂₀O₃
Bod varu: 158 °C
Bod tání: < 0 °C
Hustota: 0,880
Bod vzplanutí: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009226
CAS číslo: 115-80-0
EINECS: 204-108-0
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR