Tisk…

1,7-Octadiene ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L07659.AE L07659.AP
L07659.AEEA 1520 CZK
L07659.AE L07659.AP
1,7-Octadiene ≥97%
1,7-Octadiene

Purity: 97% CAS No.: 3710-30-3 Molecular Formula: C8H14 Formula Weight: 110.20 Flash Point: 5°(41°F) Refractive Index: 1.4220 Odour: Strong Melting Point: -70° Boiling Point: 118-120° Beilstein Reference: 605288 UN No.: 2309 MDL No.: MFCD00008668

Vzorec: C₈H₁₄
Bod varu: 118…120 °C
Bod tání: < –70 °C
Hustota: 0,735
Bod vzplanutí: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008668
CAS číslo: 3710-30-3
EINECS: 223-054-9
UN: 2309
ADR: 3,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR