Tisk…

Ethyl-2-iodobenzoate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ethyl 2-iodobenzoate, 2-Iodobenzoic acid ethyl ester

L07650.14 L07650.06
L07650.14EA 4980 CZK
L07650.14 L07650.06
Ethyl-2-iodobenzoate ≥98%
Ethyl-2-iodobenzoate

Purity: 98% CAS No.: 1829-28-3 Molecular Formula: C9H9IO2 Formula Weight: 276.08 Refractive Index: 1.5840 Boiling Point: 163-165°/23mm MDL No.: MFCD00051796

Vzorec: C₉H₉IO₂
MW: 276.07
Bod varu: 163…165 °C (23 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00051796
CAS číslo: 1829-28-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR