Tisk…

Phthalide-3-acetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L05984.14 L05984.06
L05984.14EA 1770 CZK
L05984.14 L05984.06
Phthalide-3-acetic acid ≥98%
Phthalide-3-acetic acid

Purity: 98+% CAS No.: 4743-58-2 Molecular Formula: C10H8O4 Formula Weight: 192.17 Melting Point: 148-153° Beilstein Reference: 156005 MDL No.: MFCD00051729

Vzorec: C₁₀H₈O₄
MW: 192.17 g/mol
Bod tání: 148…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00051729
CAS číslo: 4743-58-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR