Tisk…

trans-2-Decenal 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: trans-2-Decenal, (E)-2-Decenal, (E)-Dec-2-enal

L05901.06 L05901.14
L05901.06EA 1920 CZK
L05901.06 L05901.14
trans-2-Decenal 95%
trans-2-Decenal

Purity: 95% CAS No.: 3913-81-3 Molecular Formula: C10H18O Formula Weight: 154.25 Storage Temperature: 2°C to 8°C Flash Point: 93°(199°F) Refractive Index: 1.4565 Boiling Point: 117°/17mm Beilstein Reference: 1704445 MDL No.: MFCD00014679

Vzorec: C₁₀H₁₈O
MW: 154.25
Bod varu: 117 °C (17 mmHg)
Hustota: 0.840
Bod vzplanutí: 93 °C (199 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00014679
CAS číslo: 3913-81-3
EINECS: 223-474-2
UN: 3082
ADR: 9 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR