Tisk…

2-(2-Aminoethyl)pyridine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L05627.03 L05627.06 L05627.14
L05627.03EA 1060 CZK
L05627.03 L05627.06 L05627.14
2-(2-Aminoethyl)pyridine 98%
2-(2-Aminoethyl)pyridine

Purity: 98% CAS No.: 2706-56-1 Molecular Formula: C7H10N2 Formula Weight: 122.17 Flash Point: 100°(212°F) Refractive Index: 1.5360 Odour: Amine-like Boiling Point: 208° Beilstein Reference: 111208 UN No.: 2735 MDL No.: MFCD00006367

Vzorec: C₇H₁₀N₂
Bod varu: 208 °C
Hustota: 1,020
Bod vzplanutí: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006367
CAS číslo: 2706-56-1
EINECS: 220-295-1
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR