Tisk…

Citraconic anhydride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Methylfuran-2,5-dione

L05238.14 L05238.22 L05238.36
L05238.14EA 930 CZK
L05238.14 L05238.22 L05238.36
Citraconic anhydride ≥98%
Citraconic anhydride

Purity: 98% CAS No.: 616-02-4 Molecular Formula: C5H4O3 Formula Weight: 112.08 Flash Point: 101°(213°F) Refractive Index: 1.4710 Melting Point: 7-8° Boiling Point: 213-214° Beilstein Reference: 1835 UN No.: 2810 MDL No.: MFCD00005522

Vzorec: C₅H₄O₃
MW: 112.08
Bod varu: 213…214 °C
Bod tání: 7…8 °C
Hustota: 1.243
Bod vzplanutí: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005522
CAS číslo: 616-02-4
EINECS: 210-459-0
UN: 2810
ADR: 6.1 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR