Tisk…

Allyl phenyl sulphide ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L04771.09
L04771.09EA 1900 CZK
L04771.09
Allyl phenyl sulphide ≥97%
Allyl phenyl sulphide

Purity: 97% CAS No.: 5296-64-0 Molecular Formula: C9H10S Formula Weight: 150.24 Flash Point: 98°(208°F) Refractive Index: 1.5740 Odour: Unpleasant Boiling Point: 223-225° Beilstein Reference: 1856940 MDL No.: MFCD00014957

Vzorec: C₉H₁₀S
Bod varu: 223…225 °C
Hustota: 1,024
Bod vzplanutí: >98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014957
CAS číslo: 5296-64-0
EINECS: 226-142-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR