Tisk…

Citraconic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Methyl-2-butenedioic acid

L04178.09 L04178.18
L04178.09EA 1100 CZK
L04178.09 L04178.18
Citraconic acid ≥98%
Citraconic acid

Purity: 98+% CAS No.: 498-23-7 Molecular Formula: C5H6O4 Formula Weight: 130.10 Melting Point: 88-91° Beilstein Reference: 1722679 Merck Reference: 14,2321 MDL No.: MFCD00078086

Vzorec: C₅H₆O₄
Bod varu: 336 °C (1013 hPa)
Bod tání: 88…91 °C
Hustota: 1,620
Storage Temperature: Ambient
MDL Number: MFCD00078086
CAS číslo: 498-23-7
EINECS: 207-858-7
Merck Index: 14,2321

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR