Tisk…

α-Fluorophenylacetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L01745.04 L01745.MC L01745.ME
L01745.04EA 6460 CZK
L01745.04 L01745.MC L01745.ME
α-Fluorophenylacetic acid ≥98%
α-Fluorophenylacetic acid

Purity: 98% CAS No.: 1578-63-8 Molecular Formula: C6H5CHFCO2H Formula Weight: 154.14 Melting Point: 78-82° Beilstein Reference: 2087378 UN No.: 2811 MDL No.: MFCD00004221

Vzorec: C₈H₇FO₂
Bod tání: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004221
CAS číslo: 1578-63-8
EINECS: 216-420-4
UN: 2811
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR