Tisk…

3-Bromo-1,2,4,5-tetramethylbenzene 99

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Bromo-2,3,5,6-tetramethylbenzene

L01465.06 L01465.14
L01465.06EA 2430 CZK
L01465.06 L01465.14
3-Bromo-1,2,4,5-tetramethylbenzene 99
3-Bromo-1,2,4,5-tetramethylbenzene

Purity: 99% CAS No.: 1646-53-3 Molecular Formula: C10H13Br Formula Weight: 213.12 Melting Point: 58-62° Boiling Point: 115-120°/5mm MDL No.: MFCD00000070

Vzorec: C₁₀H₁₃Br
MW: 213.12
Bod varu: 115…120 °C (5 mmHg)
Bod tání: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000070
CAS číslo: 1646-53-3
EINECS: 216-707-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR