Tisk…

Piperidin-3-ol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L01333.04 L01333.09 L01333.18
L01333.04EA 704 CZK
L01333.04 L01333.09 L01333.18
Piperidin-3-ol ≥98%
Piperidin-3-ol

Purity: 98+% CAS No.: 6859-99-0 Molecular Formula: C5H11NO Formula Weight: 101.15 Storage Temperature: 2°C to 8°C Flash Point: 100°(212°F) Odour: Amine-like pH: 11.5-11.7 Melting Point: 56-60° Boiling Point: 67-69°/2mm Beilstein Reference: 102696 UN No.: 3263 MDL No.: MFCD00014591

Vzorec: C₅H₁₁NO
Bod varu: 67…69 °C (2 mmHg)
Bod tání: 56…60 °C
Bod vzplanutí: >100 °C (212 °F)
MDL Number: MFCD00014591
CAS číslo: 6859-99-0
EINECS: 229-957-4
UN: 3263
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR