Tisk…

Triphenylene 98

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 9,10-Benzophenanthrene, Isochrysene

L00955.06
L00955.06EA 6870 CZK
L00955.06
Triphenylene 98
Triphenylene

Purity: 98% CAS No.: 217-59-4 Molecular Formula: C18H12 Formula Weight: 228.30 Melting Point: 194-199° Boiling Point: 438° Merck Reference: 14,9740 MDL No.: MFCD00001108

Vzorec: C₁₈H₁₂
MW: 228.29
Bod varu: 438 °C
Bod tání: 194…199 °C
Hustota: 1.302
Storage Temperature: Ambient
MDL Number: MFCD00001108
CAS číslo: 217-59-4
EINECS: 205-922-9
UN: 3077
ADR: 9 III
Merck Index: 12,09870

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR